Anti-PIK3R4 Mouse Monoclonal Antibody [clone: 2A6]

Supplier: Abnova
H00030849-M05
10596-996EA 335 USD
10596-996
Anti-PIK3R4 Mouse Monoclonal Antibody [clone: 2A6]
Antibodies
Type: Primary
Antigen: PIK3R4
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR