Anti-EHD3 Mouse Monoclonal Antibody [clone: 4B7]

Supplier: Abnova
H00030845-M01
10584-772EA 335 USD
10584-772
Anti-EHD3 Mouse Monoclonal Antibody [clone: 4B7]
Antibodies
Type: Primary
Antigen: EHD3
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR