Anti-SNX11 Mouse Monoclonal Antibody [clone: 2G1]

Supplier: Abnova
H00029916-M05
10602-336EA 335 USD
10602-336
Anti-SNX11 Mouse Monoclonal Antibody [clone: 2G1]
Antibodies
Type: Primary
Antigen: SNX11
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR