Anti-FAM216A Mouse Monoclonal Antibody [clone: 2G4-1A1]

Supplier: Abnova
H00029902-M01
10589-540EA 335 USD
10589-540
Anti-FAM216A Mouse Monoclonal Antibody [clone: 2G4-1A1]
Antibodies
Type: Primary
Antigen: FAM216A (family with sequence similarity 216, member A)
Clonality: Monoclonal
Clone: 2G4-1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR