Anti-CLEC2D Mouse Monoclonal Antibody [clone: 4C7]

Supplier: Abnova
H00029121-M01
10591-904EA 335 USD
10591-904
Anti-CLEC2D Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
Type: Primary
Antigen: CLEC2D
Clonality: Monoclonal
Clone: 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR