Anti-FHOD1 Mouse Monoclonal Antibody [clone: 3F7]

Supplier: Abnova
H00029109-M02
10586-060EA 335 USD
10586-060
Anti-FHOD1 Mouse Monoclonal Antibody [clone: 3F7]
Antibodies
Type: Primary
Antigen: FHOD1 (formin homology 2 domain containing 1)
Clonality: Monoclonal
Clone: 3F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR