Anti-FLVCR1 Mouse Monoclonal Antibody [clone: 4B2]

Supplier: Abnova
H00028982-M05
10586-272EA 335 USD
10586-272
Anti-FLVCR1 Mouse Monoclonal Antibody [clone: 4B2]
Antibodies
Type: Primary
Antigen: FLVCR1 (feline leukemia virus subgroup C cellular receptor 1)
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR