Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 4G4-D5]

Supplier: Abnova
H00026119-M01
10577-200EA 335 USD
10577-200
Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 4G4-D5]
Antibodies
Type: Primary
Antigen: LDLRAP1
Clonality: Monoclonal
Clone: 4G4-D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR