Anti-TWF2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00011344-D01P
10598-580EA 376 USD
10598-580
Anti-TWF2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TWF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR