Anti-RNF13 Mouse Polyclonal Antibody

Supplier: Abnova
H00011342-A01
10570-002EA 243 USD
10570-002
Anti-RNF13 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR