Anti-EXOSC8 Mouse Monoclonal Antibody [clone: 4B3]

Supplier: Abnova
H00011340-M02
10585-420EA 335 USD
10585-420
Anti-EXOSC8 Mouse Monoclonal Antibody [clone: 4B3]
Antibodies
Type: Primary
Antigen: EXOSC8
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR