Anti-PDAP1 Mouse Monoclonal Antibody [clone: 3B10]

Supplier: Abnova
H00011333-M05
10596-372EA 335 USD
10596-372
Anti-PDAP1 Mouse Monoclonal Antibody [clone: 3B10]
Antibodies
Type: Primary
Antigen: PDAP1
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR