Anti-STK38 Mouse Monoclonal Antibody [clone: 2F3]

Supplier: Abnova
H00011329-M04
10602-758EA 335 USD
10602-758
Anti-STK38 Mouse Monoclonal Antibody [clone: 2F3]
Antibodies
Type: Primary
Antigen: STK38
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR