Anti-ECD Mouse Monoclonal Antibody [clone: 1E5]

Supplier: Abnova
H00011319-M02
10589-440EA 335 USD
10589-440
Anti-ECD Mouse Monoclonal Antibody [clone: 1E5]
Antibodies
Type: Primary
Antigen: ECD
Clonality: Monoclonal
Clone: 100000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR