Anti-CD300A Mouse Polyclonal Antibody

Supplier: Abnova
H00011314-B01P
10553-708EA 335 USD
10553-708
Anti-CD300A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR