Anti-MGAT4B Mouse Monoclonal Antibody [clone: 1F4]

Supplier: Abnova
H00011282-M01
10593-310EA 335 USD
10593-310
Anti-MGAT4B Mouse Monoclonal Antibody [clone: 1F4]
Antibodies
Type: Primary
Antigen: MGAT4B
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR