Anti-POU6F2 Mouse Monoclonal Antibody [clone: 1D3]

Supplier: Abnova
H00011281-M05
10597-690EA 335 USD
10597-690
Anti-POU6F2 Mouse Monoclonal Antibody [clone: 1D3]
Antibodies
Type: Primary
Antigen: POU6F2
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR