Anti-KLF8 Mouse Monoclonal Antibody [clone: 2E10]

Supplier: Abnova
H00011279-M09
10591-208EA 335 USD
10591-208
Anti-KLF8 Mouse Monoclonal Antibody [clone: 2E10]
Antibodies
Type: Primary
Antigen: KLF8
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR