Anti-KLF12 Mouse Monoclonal Antibody [clone: 30000]

Supplier: Abnova
H00011278-M01
10591-192EA 335 USD
10591-192
Anti-KLF12 Mouse Monoclonal Antibody [clone: 30000]
Antibodies
Type: Primary
Antigen: KLF12
Clonality: Monoclonal
Clone: 30000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR