Anti-SNF8 (SNF8, ESCRT-II Complex Subunit) Mouse Polyclonal Antibody

Supplier: Abnova
H00011267-B01
10579-330EA 335 USD
10579-330
Anti-SNF8 (SNF8, ESCRT-II Complex Subunit) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNF8 (SNF8, ESCRT-II Complex Subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR