Anti-CHP1 Mouse Monoclonal Antibody [clone: S2]

Supplier: Abnova
H00011261-M02
10582-176EA 335 USD
10582-176
Anti-CHP1 Mouse Monoclonal Antibody [clone: S2]
Antibodies
Type: Primary
Antigen: CHP1
Clonality: Monoclonal
Clone: S2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR