Anti-PACSIN2 Mouse Monoclonal Antibody [clone: 3D6]

Supplier: Abnova
H00011252-M05
10595-910EA 335 USD
10595-910
Anti-PACSIN2 Mouse Monoclonal Antibody [clone: 3D6]
Antibodies
Type: Primary
Antigen: PACSIN2
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR