Anti-NXPH3 Mouse Monoclonal Antibody [clone: 4C8]

Supplier: Abnova
H00011248-M05
10595-606EA 335 USD
10595-606
Anti-NXPH3 Mouse Monoclonal Antibody [clone: 4C8]
Antibodies
Type: Primary
Antigen: NXPH3
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR