Anti-PADI2 Mouse Monoclonal Antibody [clone: 4D4]

Supplier: Abnova
H00011240-M01
10594-816EA 335 USD
10594-816
Anti-PADI2 Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
Type: Primary
Antigen: PADI2
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR