Anti-RASSF8 Mouse Monoclonal Antibody [clone: 2G1]

Supplier: Abnova
H00011228-M01
10578-222EA 335 USD
10578-222
Anti-RASSF8 Mouse Monoclonal Antibody [clone: 2G1]
Antibodies
Type: Primary
Antigen: RASSF8
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR