Anti-GALNT6 Mouse Polyclonal Antibody

Supplier: Abnova
H00011226-B01
10557-300EA 335 USD
10557-300
Anti-GALNT6 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GALNT6 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR