Anti-CHEK2 Mouse Monoclonal Antibody [clone: 3E10]

Supplier: Abnova
H00011200-M03
10553-402EA 335 USD
10553-402
Anti-CHEK2 Mouse Monoclonal Antibody [clone: 3E10]
Antibodies
Type: Primary
Antigen: CHEK2
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR