Anti-CHEK2 Mouse Monoclonal Antibody [clone: 6B8]

Supplier: Abnova
H00011200-M02A
10624-054EA 335 USD
10624-054
Anti-CHEK2 Mouse Monoclonal Antibody [clone: 6B8]
Antibodies
Type: Primary
Antigen: CHEK2
Clonality: Monoclonal
Clone: 6B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR