Anti-SUPT16H Mouse Monoclonal Antibody [clone: 4D6]

Supplier: Abnova
H00011198-M07
10602-414EA 335 USD
10602-414
Anti-SUPT16H Mouse Monoclonal Antibody [clone: 4D6]
Antibodies
Type: Primary
Antigen: SUPT16H
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR