Anti-NISCH Mouse Monoclonal Antibody [clone: 3B8]

Supplier: Abnova
H00011188-M01
10594-940EA 335 USD
10594-940
Anti-NISCH Mouse Monoclonal Antibody [clone: 3B8]
Antibodies
Type: Primary
Antigen: NISCH (nischarin)
Clonality: Monoclonal
Clone: 3B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR