Anti-PSIP1 Mouse Monoclonal Antibody [clone: 3H1]

Supplier: Abnova
H00011168-M01
10597-598EA 335 USD
10597-598
Anti-PSIP1 Mouse Monoclonal Antibody [clone: 3H1]
Antibodies
Type: Primary
Antigen: PSIP1 (PC4 and SFRS1 interacting protein 1)
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR