Anti-SOX21 Mouse Monoclonal Antibody [clone: 3H4]

Supplier: Abnova
H00011166-M02
10601-778EA 335 USD
10601-778
Anti-SOX21 Mouse Monoclonal Antibody [clone: 3H4]
Antibodies
Type: Primary
Antigen: SOX21
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR