Anti-NUDT6 Mouse Polyclonal Antibody

Supplier: Abnova
H00011162-B01P
10565-396EA 335 USD
10565-396
Anti-NUDT6 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NUDT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR