Anti-RABL2B Mouse Monoclonal Antibody [clone: 1B10]

Supplier: Abnova
H00011158-M01
10599-016EA 335 USD
10599-016
Anti-RABL2B Mouse Monoclonal Antibody [clone: 1B10]
Antibodies
Type: Primary
Antigen: RABL2B
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR