Anti-CORO1A Mouse Monoclonal Antibody [clone: 1A8]

Supplier: Abnova
H00011151-M03
10582-684EA 335 USD
10582-684
Anti-CORO1A Mouse Monoclonal Antibody [clone: 1A8]
Antibodies
Type: Primary
Antigen: CORO1A
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR