Anti-GLMN Rabbit Polyclonal Antibody

Supplier: Abnova
H00011146-D01
10587-456EA 376 USD
10587-456
Anti-GLMN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GLMN (glomulin, FKBP associated protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR