Anti-PWP1 Mouse Monoclonal Antibody [clone: 1A12]

Supplier: Abnova
H00011137-M03
10597-852EA 335 USD
10597-852
Anti-PWP1 Mouse Monoclonal Antibody [clone: 1A12]
Antibodies
Type: Primary
Antigen: PWP1
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR