Anti-ZWINT Mouse Monoclonal Antibody [clone: 6D3]

Supplier: Abnova
H00011130-M01
10606-638EA 335 USD
10606-638
Anti-ZWINT Mouse Monoclonal Antibody [clone: 6D3]
Antibodies
Type: Primary
Antigen: ZWINT
Clonality: Monoclonal
Clone: 6D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR