Anti-CD160 Rabbit Polyclonal Antibody

Supplier: Abnova
H00011126-D01
10579-016EA 376 USD
10579-016
Anti-CD160 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR