Anti-FGFR1OP Mouse Monoclonal Antibody [clone: 2B1]

Supplier: Abnova
H00011116-M01
10585-962EA 335 USD
10585-962
Anti-FGFR1OP Mouse Monoclonal Antibody [clone: 2B1]
Antibodies
Type: Primary
Antigen: FGFR1OP (FGFR1 oncogene partner)
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR