Anti-FGFR1OP Mouse Polyclonal Antibody

Supplier: Abnova
H00011116-B01P
10556-580EA 335 USD
10556-580
Anti-FGFR1OP Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGFR1OP (FGFR1 oncogene partner)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR