Anti-KATNA1 Mouse Polyclonal Antibody

Supplier: Abnova
H00011104-B02
10562-124EA 335 USD
10562-124
Anti-KATNA1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KATNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR