Anti-RPP14 (ribonuclease P/MRP 14kDa Subunit) Mouse Monoclonal Antibody [clone: 3H4]

Supplier: Abnova
H00011102-M01A
10625-042EA 335 USD
10625-042
Anti-RPP14 (ribonuclease P/MRP 14kDa Subunit) Mouse Monoclonal Antibody [clone: 3H4]
Antibodies
Type: Primary
Antigen: RPP14 (ribonuclease P/MRP 14kDa Subunit)
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR