Anti-RPP14 (ribonuclease P/MRP 14kDa Subunit) Mouse Polyclonal Antibody

Supplier: Abnova
H00011102-B01
10570-320EA 335 USD
10570-320
Anti-RPP14 (ribonuclease P/MRP 14kDa Subunit) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPP14 (ribonuclease P/MRP 14kDa Subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR