Anti-ADAMTS13 Mouse Monoclonal Antibody [clone: 4F12]

Supplier: Abnova
H00011093-M06
10576-232EA 335 USD
10576-232
Anti-ADAMTS13 Mouse Monoclonal Antibody [clone: 4F12]
Antibodies
Type: Primary
Antigen: ADAMTS13
Clonality: Monoclonal
Clone: 4F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR