Anti-DIDO1 Mouse Monoclonal Antibody [clone: 3B1]

Supplier: Abnova
H00011083-M04
10583-526EA 335 USD
10583-526
Anti-DIDO1 Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
Type: Primary
Antigen: DIDO1
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR