Anti-DIDO1 Mouse Monoclonal Antibody [clone: 3000]

Supplier: Abnova
H00011083-M01A
10623-934EA 335 USD
10623-934
Anti-DIDO1 Mouse Monoclonal Antibody [clone: 3000]
Antibodies
Type: Primary
Antigen: DIDO1
Clonality: Monoclonal
Clone: 3000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR