Anti-DNAJB4 Mouse Monoclonal Antibody [clone: 1H2-3G4]

Supplier: Abnova
H00011080-M01
10584-188EA 335 USD
10584-188
Anti-DNAJB4 Mouse Monoclonal Antibody [clone: 1H2-3G4]
Antibodies
Type: Primary
Antigen: DNAJB4
Clonality: Monoclonal
Clone: 1H2-3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR