Anti-STMN2 Mouse Monoclonal Antibody [clone: 1A7]

Supplier: Abnova
H00011075-M23
10579-794EA 335 USD
10579-794
Anti-STMN2 Mouse Monoclonal Antibody [clone: 1A7]
Antibodies
Type: Primary
Antigen: STMN2
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR