Anti-STMN2 Mouse Monoclonal Antibody [clone: 1C6]

Supplier: Abnova
H00011075-M02
10602-780EA 335 USD
10602-780
Anti-STMN2 Mouse Monoclonal Antibody [clone: 1C6]
Antibodies
Type: Primary
Antigen: STMN2
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR